Assay Details
| Gene Symbol: FAM19A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082967.2 | NP_001076436.1 | 1 - 2 | 243 | 90 | |
| GenBank mRNA | AF131851.1 | - | 1 - 2 | 209 | 90 | |
| BC039396.1 | - | 1 - 2 | 222 | 5299791 | 90 | |
| DB481230.1 | - | 1 - 2 | 90 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000407.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22007805.hg.1 | ENST00000402357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007805.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007805.hg.1 | TC22000407.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007808.hg.1 | ENST00000358295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007808.hg.1 | ENST00000402357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007808.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007808.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007808.hg.1 | TC22000407.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007804.hg.1 | ENST00000402357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007804.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007804.hg.1 | TC22000407.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000406880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000406880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000406880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169449.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169449.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169449.hg.1 | TC2200009290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169449.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169449.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169450.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169450.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169450.hg.1 | TC2200009290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169450.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169450.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000358295.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000402357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000406880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000406880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000406880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | ENST00000473898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_001082967.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009290.hg.1 | NM_015381.6 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 167237 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 235066 | - | - |

FAM19A5
Genomic Map