Assay Details
| Gene Symbol: KLHL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025010.4 | NP_079286.2 | 1 - 2 | 262 | 88 | |
| XM_005265001.3 | XP_005265058.2 | 1 - 2 | 162 | 88 | ||
| XM_005265002.4 | XP_005265059.2 | 1 - 2 | 162 | 88 | ||
| XM_005265003.3 | XP_005265060.2 | 1 - 2 | 162 | 88 | ||
| GenBank mRNA | AI356423.1 | - | 1 - 2 | 262 | 88 | |
| AK291477.1 | - | 1 - 2 | 156 | 88 | ||
| BC032620.1 | - | 1 - 2 | 74 | 5578173 | 88 | |
| BX537953.1 | - | 1 - 2 | 153 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000267.hg
Additional Information:
This assay is in the 5'UTR region of the BC032620.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03005438.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005438.hg.1 | ENST00000455924.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005438.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005438.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005438.hg.1 | TC03000267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005439.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005439.hg.1 | ENST00000455924.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005439.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005439.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005439.hg.1 | TC03000267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005442.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005442.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005442.hg.1 | TC03000267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005434.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005434.hg.1 | TC03000267.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005435.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005435.hg.1 | TC03000267.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007290.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151697.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151697.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151697.hg.1 | TC0300007290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151706.hg.1 | TC0300007290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151698.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151698.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151698.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151698.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151698.hg.1 | TC0300007290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151703.hg.1 | TC0300007290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000232766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000433449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000437353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000442272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000461084.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | ENST00000483201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | NM_025010.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151702.hg.1 | TC0300007290.hg.1 | Clariom D TC |

KLHL18
Genomic Map