| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005446.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005446.hg.1 |
ENST00000455924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005446.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005446.hg.1 |
TC03000267.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000483201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000483201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
ENST00000483201.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151710.hg.1 |
TC0300007290.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000433449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000483201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000483201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
ENST00000483201.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007290.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000232766.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000437353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000442272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
ENST00000461084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
NM_025010.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151709.hg.1 |
TC0300007290.hg.1 |
Clariom D TC |