Assay Details
| Gene Symbol: DARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018122.4 | NP_060592.2 | 14 - 15 | 2137 | 97 | |
| XM_006711427.3 | XP_006711490.1 | 13 - 14 | 1941 | 97 | ||
| GenBank mRNA | AK001376.1 | - | 10 - 11 | 1141 | 97 | |
| AK022754.1 | - | 10 - 11 | 1142 | 97 | ||
| AK290873.1 | - | 14 - 15 | 1819 | 97 | ||
| AL833633.1 | - | 14 - 15 | 1822 | 97 | ||
| BC045173.2 | - | 14 - 15 | 2116 | 5273243 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001516.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023447.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023447.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023447.hg.1 | TC01001516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023446.hg.1 | ENST00000239457.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023446.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023446.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023446.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023446.hg.1 | TC01001516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178379.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178379.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178379.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178379.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178379.hg.1 | TC0100010661.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178378.hg.1 | TC0100010661.hg.1 | Clariom D TC |

DARS2
Genomic Map