Assay Details
| Gene Symbol: DARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018122.4 | NP_060592.2 | 4 - 5 | 963 | 107 | |
| XM_006711427.3 | XP_006711490.1 | 4 - 5 | 920 | 107 | ||
| XM_011509711.2 | XP_011508013.1 | 4 - 5 | 920 | 107 | ||
| GenBank mRNA | AK290873.1 | - | 4 - 5 | 645 | 107 | |
| AL833633.1 | - | 4 - 5 | 648 | 107 | ||
| BC045173.2 | - | 4 - 5 | 942 | 5273243 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001516.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01023434.hg.1 | TC01001516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023434.hg.1 | ENST00000239457.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023434.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01023434.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178366.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178366.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178366.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100178366.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100178366.hg.1 | TC0100010661.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000361951.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000460076.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | ENST00000471476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010661.hg.1 | NM_018122.4 | Refseq |

DARS2
Genomic Map