Assay Details
| Gene Symbol: HCP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC113853.1 | - | 1 | 149 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000467369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000467369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000467369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000460889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000467369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000467369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000467369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | ENST00000541196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014102.hg.1 | NR_040662.1 | Refseq |

HCP5
Genomic Map