Assay Details
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198041.2 | NP_932158.1 | 1 - 2 | 205 | 82 | |
| GenBank mRNA | AF019632.2 | - | 1 - 2 | 169 | 82 | |
| AK294450.1 | - | 1 - 2 | 205 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001523.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF019632.2,AK294450.1,NM_198041.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04021641.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021641.hg.1 | TC04001523.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021640.hg.1 | ENST00000339154.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021640.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021640.hg.1 | TC04001523.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190955.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190955.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190955.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190955.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190955.hg.1 | TC0400012961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400190956.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190956.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190956.hg.1 | TC0400012961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |

NUDT6
Genomic Map