Assay Details
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007083.4 | NP_009014.2 | 3 - 4 | 538 | 127 | |
| NM_198041.2 | NP_932158.1 | 3 - 4 | 470 | 127 | ||
| GenBank mRNA | AF019632.2 | - | 3 - 4 | 434 | 127 | |
| AF019633.2 | - | 3 - 4 | 538 | 127 | ||
| AK291871.1 | - | 3 - 4 | 537 | 127 | ||
| AK294450.1 | - | 3 - 4 | 470 | 127 | ||
| BC009842.2 | - | 3 - 4 | 518 | 3942416 | 127 | |
| BG386740.1 | - | 3 - 4 | 516 | 127 | ||
| CB128685.1 | - | 3 - 4 | 539 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001523.hg
Additional Information:
This assay is in the 5'UTR region of the AF019632.2,AK294450.1,NM_198041.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04021627.hg.1 | ENST00000304430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021627.hg.1 | ENST00000339154.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021627.hg.1 | ENST00000502270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021627.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021627.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021627.hg.1 | TC04001523.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190940.hg.1 | TC0400012961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000304430.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000339154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000502270.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000503370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000510735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000512116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | ENST00000513517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_007083.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012961.hg.1 | NM_198041.2 | Refseq |

NUDT6
Genomic Map