Assay Details
| Gene Symbol: UNC5CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173561.2 | NP_775832.2 | 4 - 5 | 1048 | 64 | |
| GenBank mRNA | AY510109.1 | - | 3 - 4 | 949 | 64 | |
| BC035284.1 | - | 4 - 5 | 1038 | 5189247 | 64 | |
| JF681947.1 | - | 4 - 5 | 1066 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004133.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020370.hg.1 | ENST00000244565.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020370.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020370.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020370.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020370.hg.1 | TC06004133.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020369.hg.1 | ENST00000244565.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020369.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020369.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020369.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020369.hg.1 | TC06004133.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201786.hg.1 | TC0600014290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201787.hg.1 | TC0600014290.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000244565.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000373164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | ENST00000470102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014290.hg.1 | NM_173561.2 | Refseq |

UNC5CL
Genomic Map