| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020368.hg.1 |
ENST00000244565.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020368.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020368.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020368.hg.1 |
TC06004133.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
ENST00000470102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014290.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
ENST00000244565.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
ENST00000373164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
NM_173561.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201785.hg.1 |
TC0600014290.hg.1 |
Clariom D TC |