Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000784.3 | NP_000775.1 | 3 - 4 | 1076 | 68 | |
| XM_017003488.1 | XP_016858977.1 | 2 - 3 | 1628 | 68 | ||
| GenBank mRNA | AK226039.1 | - | 3 - 4 | 1077 | 68 | |
| AK290418.1 | - | 3 - 4 | 682 | 68 | ||
| AY178622.1 | - | 3 - 4 | 642 | 68 | ||
| BC040430.1 | - | 3 - 4 | 656 | 5760376 | 68 | |
| BC051851.1 | - | 3 - 4 | 654 | 5240515 | 68 | |
| M62401.1 | - | 3 - 4 | 663 | 68 | ||
| X59812.1 | - | 3 - 4 | 839 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001311.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020304.hg.1 | ENST00000258415.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020304.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020304.hg.1 | TC02001311.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020302.hg.1 | ENST00000258415.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020302.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020302.hg.1 | TC02001311.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020305.hg.1 | ENST00000258415.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020305.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020305.hg.1 | TC02001311.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177151.hg.1 | TC0200010811.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177148.hg.1 | TC0200010811.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177150.hg.1 | TC0200010811.hg.1 | Clariom D TC |

CYP27A1
Genomic Map