Assay Details
| Gene Symbol: SCUBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173050.3 | NP_766638.2 | 7 - 8 | 968 | 60 | |
| XM_005261752.1 | XP_005261809.1 | 8 - 9 | 1019 | 60 | ||
| XM_011530389.2 | XP_011528691.1 | 8 - 9 | 1013 | 60 | ||
| XM_011530390.2 | XP_011528692.1 | 7 - 8 | 898 | 60 | ||
| GenBank mRNA | AF525689.1 | - | 8 - 9 | 861 | 60 | |
| AK127719.1 | - | 7 - 8 | 929 | 60 | ||
| BC127263.1 | - | 7 - 8 | 865 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000861.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22014563.hg.1 | ENST00000360835.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014563.hg.1 | ENST00000381243.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014563.hg.1 | ENST00000434132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014563.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014563.hg.1 | TC22000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | TC2200008907.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164652.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000290460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000360835.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000449304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000461595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000480833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | ENST00000615096.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008907.hg.1 | NM_173050.3 | Refseq |

SCUBE1
Genomic Map