Assay Details
| Gene Symbol: SNAPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012437.5 | NP_036569.1 | 1 - 2 | 234 | 57 | |
| GenBank mRNA | AF086837.1 | - | 1 - 2 | 144 | 57 | |
| AK024555.1 | - | 1 - 2 | 211 | 57 | ||
| BC000761.2 | - | 1 - 2 | 163 | 2821705 | 57 | |
| BC004494.2 | - | 1 - 2 | 164 | 3830132 | 57 | |
| BI117540.1 | - | 235 | 57 | |||
| BM051828.1 | - | 1 - 2 | 220 | 57 | ||
| BT006753.1 | - | 1 - 2 | 144 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001260.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
neurotransmitter secretion
anterograde axonal transport
endosome to lysosome transport
negative regulation of neuron projection development
viral process
synaptic vesicle exocytosis
synaptic vesicle maturation
neuron projection development
synaptic vesicle fusion to presynaptic active zone membrane
lysosome localization
melanosome organization
regulation of protein binding
synaptic vesicle transport
anterograde synaptic vesicle transport
terminal button organization
autophagosome maturation
positive regulation of late endosome to lysosome transport
SNARE binding
protein binding
intracellular protein transport
neurotransmitter secretion
anterograde axonal transport
endosome to lysosome transport
negative regulation of neuron projection development
viral process
synaptic vesicle exocytosis
synaptic vesicle maturation
neuron projection development
synaptic vesicle fusion to presynaptic active zone membrane
lysosome localization
melanosome organization
regulation of protein binding
synaptic vesicle transport
anterograde synaptic vesicle transport
terminal button organization
autophagosome maturation
positive regulation of late endosome to lysosome transport
SNARE binding
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01019219.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019219.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019219.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019219.hg.1 | TC01001260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019214.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019214.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019214.hg.1 | TC01001260.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019221.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019221.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019221.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019221.hg.1 | TC01001260.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172782.hg.1 | TC0100010046.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172788.hg.1 | TC0100010046.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172791.hg.1 | TC0100010046.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000368685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000462880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000474959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | ENST00000478558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NM_012437.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052019.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010046.hg.1 | NR_052020.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 170467 | - | - |

SNAPIN
Genomic Map