| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019223.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019223.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019223.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019223.hg.1 |
TC01001260.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100172793.hg.1 |
TC0100010046.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000368685.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000462880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000474959.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
ENST00000478558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NM_012437.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052019.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010046.hg.1 |
NR_052020.1 |
Refseq |