Assay Details
| Gene Symbol: CERS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022075.4 | NP_071358.1 | 6 - 7 | 905 | 94 | |
| NM_181746.3 | NP_859530.1 | 6 - 7 | 945 | 94 | ||
| XM_011509451.2 | XP_011507753.1 | 6 - 7 | 608 | 94 | ||
| XM_011509452.2 | XP_011507754.1 | 6 - 7 | 656 | 94 | ||
| GenBank mRNA | AF177338.2 | - | 6 - 7 | 634 | 94 | |
| AF189062.4 | - | 1 - 2 | 118 | 94 | ||
| AK001105.1 | - | 5 - 6 | 574 | 94 | ||
| AK290989.1 | - | 6 - 7 | 581 | 94 | ||
| AY091458.1 | - | 6 - 7 | 727 | 94 | ||
| BC001357.2 | - | 6 - 7 | 600 | 3051086 | 94 | |
| BC010032.2 | - | 6 - 7 | 564 | 3506408 | 94 | |
| BQ219926.1 | - | 6 - 7 | 642 | 94 | ||
More Information
Set Membership: |
|

CERS2
Genomic Map