| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004274.hg.1 |
ENST00000286827.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004274.hg.1 |
ENST00000399841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004274.hg.1 |
ENST00000541036.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004274.hg.1 |
NM_003253.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004274.hg.1 |
TC21000388.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
ENST00000286827.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
ENST00000541036.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
NM_003253.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
TC2100008555.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
ENST00000286827.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
ENST00000541036.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100158774.hg.1 |
NM_003253.3 |
Refseq |