| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007460.hg.1 |
ENST00000297438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007460.hg.1 |
ENST00000451899.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007460.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007460.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007460.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007460.hg.1 |
TC08000546.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156167.hg.1 |
TC0800008183.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000297438.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000451899.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
ENST00000520659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_001126111.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008183.hg.1 |
NM_004337.2 |
Refseq |