Assay Details
| Gene Symbol: DRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018370.2 | NP_060840.2 | 1 - 2 | 598 | 97 | |
| XM_005269004.2 | XP_005269061.1 | 1 - 2 | 483 | 97 | ||
| XM_005269005.2 | XP_005269062.1 | 1 - 2 | 598 | 97 | ||
| GenBank mRNA | AK094923.1 | - | 1 - 2 | 228 | 97 | |
| AK297785.1 | - | 1 - 2 | 242 | 97 | ||
| BC018435.1 | - | 1 - 2 | 574 | 4474297 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000793.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12010036.hg.1 | ENST00000258534.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010036.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010036.hg.1 | TC12000793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010037.hg.1 | ENST00000258534.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010037.hg.1 | ENST00000544152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010037.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010037.hg.1 | TC12000793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010038.hg.1 | ENST00000258534.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010038.hg.1 | ENST00000544152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010038.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010038.hg.1 | TC12000793.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163593.hg.1 | TC1200008629.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000546729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008629.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163589.hg.1 | TC1200008629.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | ENST00000614926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163594.hg.1 | TC1200008629.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200163587.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163587.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163587.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163587.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163587.hg.1 | TC1200008629.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000258534.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000544152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000549365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | ENST00000551403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | NM_018370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200163591.hg.1 | TC1200008629.hg.1 | Clariom D TC |

DRAM1
Genomic Map