Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 12 - 13 | 1409 | 82 | |
| NM_001198778.1 | NP_001185707.1 | 12 - 13 | 1335 | 82 | ||
| NM_001198779.1 | NP_001185708.1 | 12 - 13 | 1448 | 82 | ||
| NM_001324375.1 | NP_001311304.1 | 10 - 11 | 1212 | 82 | ||
| NM_001324376.1 | NP_001311305.1 | 13 - 14 | 1569 | 82 | ||
| NM_003591.3 | NP_003582.2 | 12 - 13 | 1401 | 82 | ||
| XM_011519743.1 | XP_011518045.1 | 14 - 15 | 1563 | 82 | ||
| XM_011519744.1 | XP_011518046.1 | 13 - 14 | 1445 | 82 | ||
| XM_011519745.1 | XP_011518047.1 | 13 - 14 | 1497 | 82 | ||
| XM_011519747.1 | XP_011518049.1 | 13 - 14 | 1482 | 82 | ||
| GenBank mRNA | AF126404.1 | - | 11 - 12 | 1174 | 82 | |
| AK095217.1 | - | 12 - 13 | 1409 | 82 | ||
| AK294080.1 | - | 10 - 11 | 1160 | 82 | ||
| AK300491.1 | - | 12 - 13 | 1293 | 82 | ||
| AK310225.1 | - | 13 - 14 | 1480 | 82 | ||
| AL832733.1 | - | 12 - 13 | 1385 | 82 | ||
| BC009591.1 | - | 12 - 13 | 1384 | 4104375 | 82 | |
| BC110901.1 | - | 12 - 13 | 1389 | 6161736 | 82 | |
| U58088.1 | - | 9 - 10 | 894 | 82 | ||
| U83410.1 | - | 12 - 13 | 1320 | 82 | ||
More Information
Set Membership: |
|

CUL2
Genomic Map