Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 15 - 16 | 1769 | 132 | |
| NM_001198778.1 | NP_001185707.1 | 15 - 16 | 1695 | 132 | ||
| NM_001198779.1 | NP_001185708.1 | 15 - 16 | 1808 | 132 | ||
| NM_001324375.1 | NP_001311304.1 | 13 - 14 | 1572 | 132 | ||
| NM_001324376.1 | NP_001311305.1 | 16 - 17 | 1929 | 132 | ||
| NM_003591.3 | NP_003582.2 | 15 - 16 | 1761 | 132 | ||
| XM_011519743.1 | XP_011518045.1 | 17 - 18 | 1923 | 132 | ||
| XM_011519744.1 | XP_011518046.1 | 16 - 17 | 1805 | 132 | ||
| XM_011519745.1 | XP_011518047.1 | 16 - 17 | 1857 | 132 | ||
| XM_011519747.1 | XP_011518049.1 | 16 - 17 | 1842 | 132 | ||
| GenBank mRNA | AF126404.1 | - | 14 - 15 | 1534 | 132 | |
| AK095217.1 | - | 15 - 16 | 1769 | 132 | ||
| AK294080.1 | - | 13 - 14 | 1520 | 132 | ||
| AK300491.1 | - | 15 - 16 | 1653 | 132 | ||
| AK310225.1 | - | 16 - 17 | 1840 | 132 | ||
| AL832733.1 | - | 15 - 16 | 1745 | 132 | ||
| BC009591.1 | - | 15 - 16 | 1744 | 4104375 | 132 | |
| BC110901.1 | - | 15 - 16 | 1749 | 6161736 | 132 | |
| U58088.1 | - | 12 - 13 | 1254 | 132 | ||
| U83410.1 | - | 15 - 16 | 1680 | 132 | ||
More Information
Set Membership: |
|

CUL2
Genomic Map