Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198777.1 | NP_001185706.1 | 4 - 5 | 556 | 104 | |
| NM_001198778.1 | NP_001185707.1 | 4 - 5 | 482 | 104 | ||
| NM_001198779.1 | NP_001185708.1 | 4 - 5 | 595 | 104 | ||
| NM_003591.3 | NP_003582.2 | 4 - 5 | 548 | 104 | ||
| XM_011519743.1 | XP_011518045.1 | 6 - 7 | 710 | 104 | ||
| XM_011519744.1 | XP_011518046.1 | 5 - 6 | 592 | 104 | ||
| XM_011519745.1 | XP_011518047.1 | 5 - 6 | 644 | 104 | ||
| XM_011519747.1 | XP_011518049.1 | 5 - 6 | 629 | 104 | ||
| GenBank mRNA | AF126404.1 | - | 3 - 4 | 321 | 104 | |
| AK095217.1 | - | 4 - 5 | 556 | 104 | ||
| AK300491.1 | - | 4 - 5 | 440 | 104 | ||
| AL832733.1 | - | 4 - 5 | 532 | 104 | ||
| BC009591.1 | - | 4 - 5 | 531 | 4104375 | 104 | |
| BC110901.1 | - | 4 - 5 | 536 | 6161736 | 104 | |
| DC401811.1 | - | 4 - 5 | 483 | 104 | ||
More Information
Set Membership: |
|

CUL2
Genomic Map