Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519747.1 | XP_011518049.1 | 2 - 3 | 281 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011519747.1 transcripts.
Set Membership: |
|

CUL2
Genomic Map