Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018644.3 | NP_061114.2 | 1 - 2 | 114 | 118 | |
| NM_054025.2 | NP_473366.1 | 1 - 2 | 114 | 118 | ||
| XM_005271506.3 | XP_005271563.1 | 1 - 2 | 182 | 118 | ||
| GenBank mRNA | BC010466.1 | - | 1 - 2 | 114 | 4157589 | 118 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002475.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC010466.1,NM_018644.3,NM_054025.2,XM_005271506.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | ENST00000312527.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | ENST00000392580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029692.hg.1 | TC11002475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | ENST00000312527.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | ENST00000392580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | ENST00000537389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029691.hg.1 | TC11002475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029695.hg.1 | ENST00000312527.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029695.hg.1 | ENST00000392580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029695.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029695.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029695.hg.1 | TC11002475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029689.hg.1 | ENST00000312527.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029689.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029689.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029689.hg.1 | TC11002475.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199077.hg.1 | TC1100012930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199070.hg.1 | TC1100012930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199072.hg.1 | TC1100012930.hg.1 | Clariom D TC |

B3GAT1
Genomic Map