Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018644.3 | NP_061114.2 | 3 - 4 | 378 | 100 | |
| NM_054025.2 | NP_473366.1 | 2 - 3 | 510 | 100 | ||
| XM_005271506.3 | XP_005271563.1 | 3 - 4 | 429 | 100 | ||
| XM_011542751.2 | XP_011541053.1 | 4 - 5 | 641 | 100 | ||
| XM_011542753.2 | XP_011541055.1 | 4 - 5 | 644 | 100 | ||
| XM_017017550.1 | XP_016873039.1 | 2 - 3 | 266 | 100 | ||
| XM_017017551.1 | XP_016873040.1 | 3 - 4 | 2203 | 100 | ||
| GenBank mRNA | AB029396.1 | - | 3 - 4 | 292 | 100 | |
| AK299637.1 | - | 2 - 3 | 196 | 100 | ||
| BC010466.1 | - | 2 - 3 | 510 | 4157589 | 100 | |
| CR457098.1 | - | 1 - 2 | 117 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | ENST00000312527.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | ENST00000392580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | ENST00000537389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029687.hg.1 | TC11002475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | ENST00000312527.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | ENST00000392580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | ENST00000537389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029686.hg.1 | TC11002475.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199065.hg.1 | TC1100012930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012930.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000312527.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000392580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000524765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000531510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | ENST00000531778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | NM_018644.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | NM_054025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199066.hg.1 | TC1100012930.hg.1 | Clariom D TC |

B3GAT1
Genomic Map