| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011240.hg.1 |
ENST00000359120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011240.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011240.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011240.hg.1 |
TC05000795.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000504522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000504522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000504522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000506591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000506591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000506591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000503848.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000504522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000504522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000504522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000506591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000506591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000506591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000509286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
ENST00000511705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008994.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
ENST00000359120.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
ENST00000511217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
NM_152550.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500162554.hg.1 |
TC0500008994.hg.1 |
Clariom D TC |