Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152550.3 | NP_689763.3 | 7 - 8 | 1559 | 66 | |
| XM_005268380.2 | XP_005268437.1 | 7 - 8 | 1555 | 66 | ||
| XM_006714757.1 | XP_006714820.1 | 7 - 8 | 1555 | 66 | ||
| XM_011537567.2 | XP_011535869.1 | 7 - 8 | 1555 | 66 | ||
| XM_011537568.2 | XP_011535870.1 | 7 - 8 | 1549 | 66 | ||
| XM_011537569.2 | XP_011535871.1 | 7 - 8 | 1555 | 66 | ||
| XM_017009136.1 | XP_016864625.1 | 7 - 8 | 1549 | 66 | ||
| GenBank mRNA | AK074234.1 | - | 7 - 8 | 1543 | 66 | |
| AK292576.1 | - | 7 - 8 | 1544 | 66 | ||
| AL833297.1 | - | 6 - 7 | 1373 | 66 | ||
| BC125106.1 | - | 7 - 8 | 1516 | 40118357 | 66 | |
| BC125107.1 | - | 7 - 8 | 1516 | 40118360 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000795.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05011250.hg.1 | ENST00000359120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011250.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011250.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011250.hg.1 | ENST00000513503.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011250.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011250.hg.1 | TC05000795.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011254.hg.1 | ENST00000359120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011254.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011254.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011254.hg.1 | ENST00000513503.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011254.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011254.hg.1 | TC05000795.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011251.hg.1 | ENST00000359120.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011251.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011251.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011251.hg.1 | ENST00000513503.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011251.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011251.hg.1 | TC05000795.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162565.hg.1 | TC0500008994.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162563.hg.1 | TC0500008994.hg.1 | Clariom D TC |

SH3RF2
Genomic Map