Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164595.1 | NP_001158067.1 | 1 - 2 | 661 | 163 | |
| GenBank mRNA | AK294535.1 | - | 1 - 2 | 124 | 163 | |
| AW297403.1 | - | 1 - 2 | 201 | 163 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000313.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12003984.hg.1 | ENST00000402685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003984.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003984.hg.1 | TC12000313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003985.hg.1 | ENST00000402685.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003985.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003985.hg.1 | TC12000313.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200151852.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151852.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151852.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151852.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151852.hg.1 | TC1200007365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200151853.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151853.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151853.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200151853.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200151853.hg.1 | TC1200007365.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000298919.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000402685.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000539469.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | ENST00000548316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_001164595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007365.hg.1 | NM_013377.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 176652 | - | - |

PDZRN4
Genomic Map