Assay Details
| Gene Symbol: EIF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003750.2 | NP_003741.1 | 20 - 21 | 3871 | 77 | |
| GenBank mRNA | AK302575.1 | - | 20 - 21 | 3816 | 77 | |
| BC020285.1 | - | 2 - 3 | 291 | 77 | ||
| BC114429.1 | - | 20 - 21 | 3779 | 40027607 | 77 | |
| D50929.1 | - | 20 - 21 | 3853 | 77 | ||
| JX312508.1 | - | 20 - 21 | 3872 | 77 | ||
| U58046.1 | - | 20 - 21 | 3823 | 77 | ||
| U78311.1 | - | 20 - 21 | 3838 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001699.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
formation of translation preinitiation complex
formation of cytoplasmic translation initiation complex
translational initiation
regulation of translational initiation
IRES-dependent viral translational initiation
viral translational termination-reinitiation
translation initiation factor activity
structural molecule activity
protein binding
poly(A) RNA binding
formation of translation preinitiation complex
formation of cytoplasmic translation initiation complex
translational initiation
regulation of translational initiation
IRES-dependent viral translational initiation
viral translational termination-reinitiation
translation initiation factor activity
structural molecule activity
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022361.hg.1 | ENST00000369144.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022361.hg.1 | ENST00000541549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022361.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022361.hg.1 | TC10001699.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000462527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011988.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | ENST00000369144.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | ENST00000541549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | NM_003750.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000182023.hg.1 | TC1000011988.hg.1 | Clariom D TC |

EIF3A
Genomic Map