Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000784.3 | NP_000775.1 | 1 - 2 | 685 | 80 | |
| GenBank mRNA | AK226039.1 | - | 1 - 2 | 686 | 80 | |
| AK290418.1 | - | 1 - 2 | 291 | 80 | ||
| AY178622.1 | - | 1 - 2 | 251 | 80 | ||
| BC040430.1 | - | 1 - 2 | 265 | 5760376 | 80 | |
| BC051851.1 | - | 1 - 2 | 263 | 5240515 | 80 | |
| M62401.1 | - | 1 - 2 | 272 | 80 | ||
| X59812.1 | - | 1 - 2 | 448 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001311.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020293.hg.1 | ENST00000258415.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020293.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020293.hg.1 | TC02001311.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020297.hg.1 | ENST00000258415.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020297.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020297.hg.1 | TC02001311.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020299.hg.1 | ENST00000258415.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020299.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020299.hg.1 | TC02001311.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010811.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | ENST00000411688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177145.hg.1 | TC0200010811.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000445971.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000466602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177143.hg.1 | TC0200010811.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | ENST00000258415.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | ENST00000494263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | NM_000784.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177138.hg.1 | TC0200010811.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 180202 | - | - |

CYP27A1
Genomic Map