Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029316.hg.1 |
TC02001831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029316.hg.1 |
ENST00000304421.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029316.hg.1 |
ENST00000346173.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029316.hg.1 |
ENST00000406846.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029316.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029316.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
ENST00000469138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012550.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000304421.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000406846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000419927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
ENST00000454032.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
NM_000145.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
NM_181446.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191548.hg.1 |
TC0200012550.hg.1 |
Clariom D TC |