Assay Details
| Gene Symbol: GPATCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018025.2 | NP_060495.2 | 5 - 6 | 867 | 95 | |
| NR_135270.1 | 5 - 6 | 867 | 95 | |||
| XM_006723255.3 | XP_006723318.1 | 5 - 6 | 582 | 95 | ||
| GenBank mRNA | AF434677.1 | - | 5 - 6 | 553 | 95 | |
| AK096005.1 | - | 5 - 6 | 561 | 95 | ||
| AK292503.1 | - | 5 - 6 | 561 | 95 | ||
| AK315712.1 | - | 5 - 6 | 579 | 95 | ||
| AL834465.1 | - | 5 - 6 | 558 | 95 | ||
| BC006108.1 | - | 5 - 6 | 867 | 3029084 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000432.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19005626.hg.1 | ENST00000170564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005626.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005626.hg.1 | TC19000432.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005625.hg.1 | ENST00000170564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005625.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005625.hg.1 | TC19000432.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000590062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | ENST00000592262.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007763.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157021.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157021.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157021.hg.1 | TC1900007763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900157021.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157021.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | NM_018025.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | TC1900007763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | ENST00000170564.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | ENST00000592165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900157022.hg.1 | NM_018025.2 | Refseq |

GPATCH1
Genomic Map