Assay Details
| Gene Symbol: SPTLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004863.3 | NP_004854.1 | 2 - 3 | 515 | 112 | |
| XM_011537384.2 | XP_011535686.1 | 2 - 3 | 411 | 112 | ||
| GenBank mRNA | AB011098.1 | - | 2 - 3 | 515 | 112 | |
| BC005123.2 | - | 2 - 3 | 416 | 3688136 | 112 | |
| U15555.1 | - | 1 - 2 | 151 | 112 | ||
| Y08686.1 | - | 2 - 3 | 375 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001348.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015342.hg.1 | ENST00000216484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015342.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015342.hg.1 | TC14001348.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000556607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009787.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | ENST00000216484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | ENST00000554901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | ENST00000557566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | NM_004863.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172289.hg.1 | TC1400009787.hg.1 | Clariom D TC |

SPTLC2
Genomic Map