| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007169.hg.1 |
ENST00000203664.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007169.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14007169.hg.1 |
TC14000582.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
ENST00000553723.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008054.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400157072.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400157072.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400157072.hg.1 |
ENST00000203664.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400157072.hg.1 |
NM_023112.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400157072.hg.1 |
TC1400008054.hg.1 |
Clariom D TC |