| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000256246.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000518257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
ENST00000523186.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010045.hg.1 |
NM_031271.3 |
Refseq |