Assay Details
| Gene Symbol: MDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014641.2 | NP_055456.2 | 8 - 9 | 3457 | 82 | |
| XM_005249492.2 | XP_005249549.1 | 8 - 9 | 3361 | 82 | ||
| XM_005249493.3 | XP_005249550.1 | 8 - 9 | 3358 | 82 | ||
| XM_005249494.4 | XP_005249551.1 | 8 - 9 | 3760 | 82 | ||
| XM_011515001.2 | XP_011513303.1 | 8 - 9 | 3188 | 82 | ||
| XM_011515003.2 | XP_011513305.1 | 8 - 9 | 3043 | 82 | ||
| XM_011515004.2 | XP_011513306.1 | 9 - 10 | 3072 | 82 | ||
| XM_017011519.1 | XP_016867008.1 | 8 - 9 | 3140 | 82 | ||
| XM_017011520.1 | XP_016867009.1 | 8 - 9 | 3056 | 82 | ||
| GenBank mRNA | BC152556.1 | - | 7 - 8 | 3030 | 9007258 | 82 |
| D79992.1 | - | 7 - 8 | 3030 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001507.hg.1, TC6_apd_hap1000077.hg.1, TC6_cox_hap2000141.hg.1, TC6_dbb_hap3000130.hg.1, TC6_mann_hap4000119.hg.1, TC6_mcf_hap5000120.hg.1, TC6_qbl_hap6000131.hg.1, TC6_ssto_hap7000121.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000494654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000494654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000494654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176739.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176739.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176739.hg.1 | TC0600011379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176744.hg.1 | TC0600011379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176743.hg.1 | TC0600011379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000416571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000417033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000422266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000425072.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000489540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000494654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000494654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000494654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011379.hg.1 | ENST00000613547.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176746.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176746.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176746.hg.1 | ENST00000376406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176746.hg.1 | ENST00000492462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600176746.hg.1 | TC0600011379.hg.1 | Clariom D TC |

MDC1
Genomic Map