| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09014004.hg.1 |
ENST00000259608.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09014004.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09014004.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09014004.hg.1 |
TC09001055.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009946.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000259608.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000474403.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000486859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
ENST00000618781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
NM_014450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168368.hg.1 |
TC0900009946.hg.1 |
Clariom D TC |