| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
ENST00000251076.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
ENST00000436119.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
ENST00000449909.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
ENST00000543779.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
NM_001174116.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
NM_001174117.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
NM_015263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013272.hg.1 |
TC15001441.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000251076.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000449909.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000543779.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000559059.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000559769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000560891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
NM_001174116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
NM_001174117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
NM_015263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
TC1500009429.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000251076.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000449909.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000543779.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000559059.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000559769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
ENST00000560891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
NM_001174116.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
NM_001174117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169258.hg.1 |
NM_015263.3 |
Refseq |