| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014657.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014657.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014657.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014657.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014658.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014658.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014658.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014658.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014663.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014663.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014663.hg.1 |
ENST00000540422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014663.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014663.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014669.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014669.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014669.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014669.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164780.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164778.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164783.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164786.hg.1 |
NM_014351.3 |
Refseq |