| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014655.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014655.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014655.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014655.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014654.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014654.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014654.hg.1 |
ENST00000540422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014654.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014654.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014656.hg.1 |
ENST00000249130.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014656.hg.1 |
ENST00000330884.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014656.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014656.hg.1 |
TC22000864.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164775.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164773.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
TC2200008920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164776.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000330884.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000422525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000432404.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
ENST00000475131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008920.hg.1 |
NM_014351.3 |
Refseq |