Assay Details
| Gene Symbol: LHX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518522.2 | XP_011516824.1 | 1 - 2 | 506 | 84 | |
| GenBank mRNA | DA799524.1 | - | 3 - 4 | 486 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011518522.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cerebral cortex radially oriented cell migration
cerebral cortex tangential migration
cerebral cortex GABAergic interneuron migration
cell maturation
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
cerebral cortex radially oriented cell migration
cerebral cortex tangential migration
cerebral cortex GABAergic interneuron migration
cell maturation
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000340587.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000373755.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000394319.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000464484.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000464484.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000464484.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000464484.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000464484.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000482062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000482062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000482062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000482062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000482062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000482062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000541397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000558672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000558672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000558672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000559895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | ENST00000560485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_001242335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_014368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011431.hg.1 | NM_199160.3 | Refseq |

LHX6
Genomic Map