Assay Details
| Gene Symbol: SWT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105518.1 | NP_001098988.1 | 7 - 8 | 1250 | 178 | |
| NM_017673.6 | NP_060143.4 | 7 - 8 | 1301 | 178 | ||
| XM_017001527.1 | XP_016857016.1 | 7 - 8 | 1188 | 178 | ||
| GenBank mRNA | AF288392.1 | - | 6 - 7 | 1147 | 178 | |
| AK127980.1 | - | 7 - 8 | 1188 | 178 | ||
| AK291443.1 | - | 7 - 8 | 1167 | 178 | ||
| AK315802.1 | - | 7 - 8 | 1268 | 178 | ||
| BC030781.2 | - | 7 - 8 | 1303 | 4821611 | 178 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001596.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01024743.hg.1 | ENST00000367500.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024743.hg.1 | ENST00000367501.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024743.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024743.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024743.hg.1 | TC01001596.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180177.hg.1 | TC0100010905.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |

SWT1
Genomic Map