| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015461.hg.1 |
ENST00000270509.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015461.hg.1 |
ENST00000341066.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015461.hg.1 |
NM_032447.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015461.hg.1 |
TC19001126.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
ENST00000270509.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
ENST00000600128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
ENST00000601739.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
NM_032447.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
TC1900009517.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
ENST00000270509.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
ENST00000600128.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
ENST00000601739.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176531.hg.1 |
NM_032447.4 |
Refseq |