Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544209.2 | XP_011542511.1 | 1 - 2 | 267 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011544209.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000295033.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000355586.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366697.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000366698.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000456946.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000457345.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000479800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | ENST00000520264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001024940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_001134855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017613.hg.1 | NM_016102.3 | Refseq |

TRIM17
Genomic Map