| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03031808.hg.1 |
ENST00000322623.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03031808.hg.1 |
ENST00000417360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03031808.hg.1 |
ENST00000464873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03031808.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03031808.hg.1 |
TC03001751.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000410695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000480616.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000480616.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300208781.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300208781.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300208781.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300208781.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300208781.hg.1 |
TC0300014041.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000322623.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000410695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000472125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478243.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478243.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478243.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000478892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000480616.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000480616.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000582176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
ENST00000585057.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300014041.hg.1 |
NM_003707.2 |
Refseq |