Assay Details
| Gene Symbol: ATXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311649.1 | - | 1 - 2 | 166 | 117 | |
| BC011026.1 | - | 1 - 2 | 199 | 117 | ||
| BF670717.1 | - | 194 | 117 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ATXN1
Genomic Map