| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014190.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014190.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014190.hg.1 |
TC08001099.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014191.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014191.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014191.hg.1 |
TC08001099.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800168102.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800168102.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800168102.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800168102.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800168102.hg.1 |
TC0800010002.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800168103.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800168103.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800168103.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800168103.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800168103.hg.1 |
TC0800010002.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240100.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
ENST00000240101.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_001394.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010002.hg.1 |
NM_057158.3 |
Refseq |