Assay Details
Gene Symbol: CPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278541.1 | NP_001265470.1 | 1 - 2 | 142 | 118 | |
NM_001872.4 | NP_001863.3 | 1 - 2 | 142 | 118 | ||
GenBank mRNA | AK290829.1 | - | 1 - 2 | 90 | 118 | |
AU100156.1 | - | 1 - 2 | 142 | 118 | ||
BC007057.1 | - | 1 - 2 | 99 | 3934520 | 118 | |
BT006936.1 | - | 1 - 2 | 75 | 118 | ||
M75106.1 | - | 1 - 2 | 94 | 118 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000641.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of extracellular matrix constituent secretion
proteolysis
blood coagulation
response to heat
negative regulation of plasminogen activation
response to drug
fibrinolysis
negative regulation of fibrinolysis
cellular response to glucose stimulus
liver regeneration
negative regulation of hepatocyte proliferation
metallocarboxypeptidase activity
zinc ion binding
positive regulation of extracellular matrix constituent secretion
proteolysis
blood coagulation
response to heat
negative regulation of plasminogen activation
response to drug
fibrinolysis
negative regulation of fibrinolysis
cellular response to glucose stimulus
liver regeneration
negative regulation of hepatocyte proliferation
metallocarboxypeptidase activity
zinc ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13007280.hg.1 | ENST00000181383.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007280.hg.1 | ENST00000439329.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007280.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007280.hg.1 | TC13000641.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007283.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007283.hg.1 | TC13000641.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13007281.hg.1 | ENST00000181383.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007281.hg.1 | ENST00000439329.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007281.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007281.hg.1 | TC13000641.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159637.hg.1 | TC1300008838.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | ENST00000181383.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159636.hg.1 | TC1300008838.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | ENST00000439329.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | NM_001278541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | NM_001872.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159640.hg.1 | TC1300008838.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 188436 | - | - |
