| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13007273.hg.1 |
ENST00000181383.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13007273.hg.1 |
ENST00000439329.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13007273.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13007273.hg.1 |
TC13000641.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300159629.hg.1 |
TC1300008838.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000181383.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
ENST00000439329.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001278541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008838.hg.1 |
NM_001872.4 |
Refseq |