Assay Details
| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278541.1 | NP_001265470.1 | 6 - 7 | 656 | 132 | |
| GenBank mRNA | AB011969.1 | - | 6 - 7 | 608 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of extracellular matrix constituent secretion
proteolysis
blood coagulation
response to heat
negative regulation of plasminogen activation
response to drug
fibrinolysis
negative regulation of fibrinolysis
cellular response to glucose stimulus
liver regeneration
negative regulation of hepatocyte proliferation
metallocarboxypeptidase activity
zinc ion binding
positive regulation of extracellular matrix constituent secretion
proteolysis
blood coagulation
response to heat
negative regulation of plasminogen activation
response to drug
fibrinolysis
negative regulation of fibrinolysis
cellular response to glucose stimulus
liver regeneration
negative regulation of hepatocyte proliferation
metallocarboxypeptidase activity
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159630.hg.1 | TC1300008838.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000181383.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | ENST00000439329.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001278541.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008838.hg.1 | NM_001872.4 | Refseq |

CPB2
Genomic Map