Assay Details
| Gene Symbol: RALBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006788.3 | NP_006779.1 | 3 - 4 | 913 | 67 | |
| GenBank mRNA | AB209924.1 | - | 1 - 2 | 120 | 67 | |
| AK300845.1 | - | 2 - 3 | 752 | 67 | ||
| AY517554.1 | - | 1 - 2 | 179 | 67 | ||
| BC013126.1 | - | 3 - 4 | 775 | 3049077 | 67 | |
| L42542.1 | - | 3 - 4 | 921 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000046.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
transport
endocytosis
chemotaxis
signal transduction
small GTPase mediated signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
transmembrane transport
GTPase activator activity
protein binding
ATPase activity
Ral GTPase binding
transmembrane transporter activity
ATPase activity, coupled to movement of substances
Rac GTPase binding
transport
endocytosis
chemotaxis
signal transduction
small GTPase mediated signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
transmembrane transport
GTPase activator activity
protein binding
ATPase activity
Ral GTPase binding
transmembrane transporter activity
ATPase activity, coupled to movement of substances
Rac GTPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000497.hg.1 | ENST00000019317.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000497.hg.1 | ENST00000383432.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000497.hg.1 | ENST00000458039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000497.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000497.hg.1 | TC18000046.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008612.hg.1 | ENST00000453252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | TC1800006639.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145413.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | TC1800006639.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145414.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008612.hg.1 | ENST00000453252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | TC1800006639.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145415.hg.1 | NM_006788.3 | Refseq |

RALBP1
Genomic Map